Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydrocyclopenta[b]indole 97.0+%, TCI America™
SDP

Catalog No. T31165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T31165G Supplier TCI America Supplier No. T31165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2047-91-8
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00225387
InChI Key HZDXFZHFEASSBM-UHFFFAOYSA-N
PubChem CID 270305
IUPAC Name 1,2,3,4-tetrahydrocyclopenta[b]indole
SMILES C1CC2=C(C1)NC3=CC=CC=C23

Specifications

Melting Point 105°C
Quantity 5 g
Formula Weight 157.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydrocyclopenta[b]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.