accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 20474-15-1

IUPAC Name: 9,9-diphenyl-9,10-dihydroacridine
Synonyms: 9,9-diphenyl-10H-acridine
Molecular Weight (g/mol): 333.43
Molecular Formula: C25H19N
InChi Key: HWTHOPMRUCFPBX-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C(C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results