Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Diphenyl-9,10-dihydroacridine 98.0+%, TCI America™
SDP

Catalog No. D5332200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5332200MG 200 mg
D53321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5332200MG Supplier TCI America Supplier No. D5332200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20474-15-1
Molecular Formula C25H19N
Molecular Weight (g/mol) 333.43
MDL Number MFCD28127357
InChI Key HWTHOPMRUCFPBX-UHFFFAOYSA-N
Synonym 9,10-Dihydro-9,9-diphenylacridine
PubChem CID 13293432
IUPAC Name 9,9-diphenyl-9,10-dihydroacridine
SMILES N1C2=CC=CC=C2C(C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 252°C
Color White-Yellow
Quantity 200 mg
Formula Weight 333.43
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Diphenyl-9,10-dihydroacridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.