CAS RN 205-95-8
CAS RN 205-95-8
IUPAC Name:
1-azapentacyclo[10.6.1.0²,⁷.0⁸,¹⁹.0¹³,¹⁸]nonadeca-2,4,6,8,10,12(19),13(18),14,16-nonaene
Synonyms:
1-azapentacyclo[10.6.1.0²,⁷.0⁸,¹⁹.0¹³,¹⁸]nonadeca-2,4,6,8,10,12(19),13(18),14,16-nonaene
Molecular Weight (g/mol):
241.29
Molecular Formula:
C18H11N
InChi Key:
SZLNOBJKCVERBJ-UHFFFAOYSA-N
SMILES:
C1=CC2=C(C=C1)C1=C3N2C2=CC=CC=C2C3=CC=C1
1
–
1
of
1
results
1
Indolo[3,2,1-jk]carbazole 98.0+%, TCI America™

CAS: 205-95-8 Molecular Formula: C18H11N Molecular Weight (g/mol): 241.293 InChI Key: SZLNOBJKCVERBJ-UHFFFAOYSA-N PubChem CID: 15352087 SMILES: C1=CC=C2C(=C1)C3=C4N2C5=CC=CC=C5C4=CC=C3