Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indolo[3,2,1-jk]carbazole 98.0+%, TCI America™
SDP

Catalog No. I1061200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1061200MG 200 mg
I10611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1061200MG Supplier TCI America Supplier No. I1061200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 205-95-8
Molecular Formula C18H11N
Molecular Weight (g/mol) 241.293
InChI Key SZLNOBJKCVERBJ-UHFFFAOYSA-N
PubChem CID 15352087
SMILES C1=CC=C2C(=C1)C3=C4N2C5=CC=CC=C5C4=CC=C3

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 200 mg
Formula Weight 241.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indolo[3,2,1-jk]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.