accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205-99-2

CAS RN 205-99-2

IUPAC Name: benzo[b]fluoranthene
Synonyms: benzo(B)fluoranthene 2,3-benzfluoranthene benz(e)acephenanthrylene 2,3-benzofluoranthrene benzo(b)fluoranthene B(b)F 3,4-benzfluoranthene ccris 72 3,4-benzofluoranthene 2,3-benzofluoranthene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: FTOVXSOBNPWTSH-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC3=CC=CC=C3C3=CC=CC2=C13
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results