Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]fluoranthene 98.0+%, TCI America™
SDP

Catalog No. B2982100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2982100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2982100MG Supplier TCI America Supplier No. B2982100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 205-99-2
Molecular Formula C20H12
Molecular Weight (g/mol) 252.316
MDL Number MFCD00010582
InChI Key FTOVXSOBNPWTSH-UHFFFAOYSA-N
Synonym benzo b fluoranthene, benz e acephenanthrylene, 3,4-benzfluoranthene, 2,3-benzofluoranthene, 2,3-benzfluoranthene, 3,4-benzofluoranthene, benzo e acephenanthrylene, b b f, 2,3-benzofluoranthrene
PubChem CID 9153
ChEBI CHEBI:34565
SMILES C1=CC=C2C3=C4C(=CC=C3)C5=CC=CC=C5C4=CC2=C1

Specifications

Melting Point 166°C
Color Yellow
Boiling Point 481°C
Quantity 100 mg
UN Number 3077
Formula Weight 252.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]fluoranthene
RTECSNumber CU1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.