accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3

CAS RN 2050-01-3

IUPAC Name: 3-methylbutyl 2-methylpropanoate
Synonyms: isoamyl isobutyrate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: VFTGLSWXJMRZNB-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C(C)C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results