Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Isobutyrate 98.0+%, TCI America™
SDP

Catalog No. I067625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I067625G 25 g
I0676250G 250 g
I0676500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I067625G Supplier TCI America Supplier No. I067625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2050-01-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00053719
InChI Key VFTGLSWXJMRZNB-UHFFFAOYSA-N
Synonym isoamyl isobutyrate, isopentyl isobutyrate, isobutyric acid isoamyl ester, propanoic acid, 2-methyl-, 3-methylbutyl ester, isobutyric acid, isopentyl ester, 3-methylbutyl isobutyrate, isoamyl 2-methylpropanoate, isopentyl 2-methylpropanoate, isoamyl isobutanoate, isopentyl isobutanoate
PubChem CID 519786
ChEBI CHEBI:87537
IUPAC Name 3-methylbutyl 2-methylpropanoate
SMILES CC(C)CCOC(=O)C(C)C

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 g
UN Number 2620
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Isobutyrate
EINECSNumber (2)-0779
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.