accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2050-25-1

IUPAC Name: 2-[2-(benzyloxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(benzyloxy)ethoxy]ethanol
Molecular Weight (g/mol): 196.25
Molecular Formula: C11H16O3
InChi Key: LJVNVNLFZQFJHU-UHFFFAOYSA-N
SMILES: OCCOCCOCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)