Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monobenzyl Ether 98.0+%, TCI America™
SDP

Catalog No. D3408500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3408500G 500 g
D340825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3408500G Supplier TCI America Supplier No. D3408500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2050-25-1
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.25
MDL Number MFCD00029649
InChI Key LJVNVNLFZQFJHU-UHFFFAOYSA-N
Synonym 2-[2-(Benzyloxy)ethoxy]ethanol
PubChem CID 229722
IUPAC Name 2-[2-(benzyloxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 500 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monobenzyl Ether
EINECSNumber (7)-0272
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.