accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClCl

CAS RN 2050-76-2

IUPAC Name: 2,4-dichloronaphthalen-1-ol
Synonyms: 2,4-dichloro-1-naphthol
Molecular Weight (g/mol): 213.06
Molecular Formula: C10H6Cl2O
InChi Key: HVLJEMXDXOTWLV-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)