accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2050-87-5

IUPAC Name: bis(prop-2-en-1-yl)trisulfane
Synonyms: diallyl trisulfide allium sativum bulb extract allium sativum bulb powder
Molecular Weight (g/mol): 178.33
Molecular Formula: C6H10S3
InChi Key: UBAXRAHSPKWNCX-UHFFFAOYSA-N
SMILES: C=CCSSSCC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results