Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl trisulfide, 98.8%, MP Biomedicals™

Catalog No. ICN19375090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19375090 0.5 g
ICN19375025 25 mg
ICN19375080 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19375090 Supplier MP Biomedicals, Inc Supplier No. 0219375090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2050-87-5
Molecular Formula C6H10S3
Molecular Weight (g/mol) 178.326
InChI Key UBAXRAHSPKWNCX-UHFFFAOYSA-N
Synonym diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide
PubChem CID 16315
ChEBI CHEBI:78492
IUPAC Name 3-(prop-2-enyltrisulfanyl)prop-1-ene
SMILES C=CCSSSCC=C

Specifications

Color Colorless
Assay Percent Range 0.988
Quantity 0.5 g
Formula Weight 178.3
Physical Form Liquid
Chemical Name or Material DATS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.