CAS RN 2051-30-1
CAS RN 2051-30-1
IUPAC Name:
2,6-dimethyloctane
Synonyms:
2,6-dimethyloctane
Molecular Weight (g/mol):
142.29
Molecular Formula:
C10H22
InChi Key:
ZALHPSXXQIPKTQ-UHFFFAOYNA-N
SMILES:
CCC(C)CCCC(C)C
1
–
1
of
1
results
1
2,6-Dimethyloctane 90.0+%, TCI America™
CAS: 2051-30-1 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00042892 InChI Key: ZALHPSXXQIPKTQ-UHFFFAOYSA-N PubChem CID: 16319 IUPAC Name: 2,6-dimethyloctane SMILES: CCC(C)CCCC(C)C