Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyloctane 90.0+%, TCI America™
SDP

Catalog No. D12121ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12121ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12121ML Supplier TCI America Supplier No. D12121ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2051-30-1
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00042892
InChI Key ZALHPSXXQIPKTQ-UHFFFAOYSA-N
PubChem CID 16319
IUPAC Name 2,6-dimethyloctane
SMILES CCC(C)CCCC(C)C

Specifications

Color Colorless
Boiling Point 158°C
Quantity 1 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyloctane
EINECSNumber (2)-0010
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.