accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2051-31-2

IUPAC Name: decan-4-ol
Synonyms: 4-decanol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: DTDMYWXTWWFLGJ-UHFFFAOYNA-N
SMILES: CCCCCCC(O)CCC
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)