Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Decanol, 97+%, Thermo Scientific™

Catalog No. AAB2086606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2086606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2086606 Supplier Thermo Scientific Chemicals Supplier No. B2086606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-31-2
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00039627
InChI Key DTDMYWXTWWFLGJ-UHFFFAOYSA-N
Synonym 4-decanol, decanol-4, 4-decanol, s, 4-decyl alcohol, hexylpropylcarbinol, 1-propylheptyl alcohol, acmc-20moz4, acmc-1cf04
PubChem CID 16320
IUPAC Name decan-4-ol
SMILES CCCCCCC(CCC)O

Specifications

Density 0.828
Boiling Point 210°C to 211°C
Flash Point 82°C (179°F)
Refractive Index 1.432
Quantity 5 g
Beilstein 1735252
Formula Weight 158.29
Percent Purity ≥97%
Chemical Name or Material 4-Decanol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-117-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.