accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C(E)

CAS RN 20515-19-9

IUPAC Name: methyl (3E)-pent-3-enoate
Synonyms: methyl (3E)-pent-3-enoate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: KJALUUCEMMPKAC-ONEGZZNKSA-N
SMILES: COC(=O)C\C=C\C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results