Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-3-Pentenoate 90.0+%, TCI America™
SDP

Catalog No. P12105ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12105ML 5 mL
P121025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12105ML Supplier TCI America Supplier No. P12105ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20515-19-9
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00191576
InChI Key KJALUUCEMMPKAC-ONEGZZNKSA-N
Synonym trans-3-Pentenoic Acid Methyl Ester
PubChem CID 642274
IUPAC Name methyl (3E)-pent-3-enoate
SMILES COC(=O)C\C=C\C

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 mL
UN Number 3272
Formula Weight 114.14
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-3-Pentenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.