CAS RN 20526-97-0
IUPAC Name:
(3Z)-3-[(4-chlorophenyl)methylidene]-1,3-dihydro-2-benzofuran-1-one
Synonyms:
(3Z)-3-[(4-chlorophenyl)methylidene]-2-benzofuran-1-one
Molecular Weight (g/mol):
256.69
Molecular Formula:
C15H9ClO2
InChi Key:
OHRFHJYUEWVXBD-ZROIWOOFSA-N
SMILES:
ClC1=CC=C(\C=C2/OC(=O)C3=CC=CC=C23)C=C1
1
–
1
of
1
results
1
3-(4-Chlorobenzal)phthalide 98.0+%, TCI America™
CAS: 20526-97-0 Molecular Formula: C15H9ClO2 Molecular Weight (g/mol): 256.69 MDL Number: MFCD02683417,MFCD02683417 InChI Key: OHRFHJYUEWVXBD-ZROIWOOFSA-N Synonym: 3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one PubChem CID: 10467624 IUPAC Name: (3Z)-3-[(4-chlorophenyl)methylidene]-1,3-dihydro-2-benzofuran-1-one SMILES: ClC1=CC=C(\C=C2/OC(=O)C3=CC=CC=C23)C=C1