Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Chlorobenzal)phthalide 98.0+%, TCI America™
SDP

Catalog No. C34481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34481G Supplier TCI America Supplier No. C34481G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20526-97-0
Molecular Formula C15H9ClO2
Molecular Weight (g/mol) 256.69
MDL Number MFCD02683417,MFCD02683417
InChI Key OHRFHJYUEWVXBD-ZROIWOOFSA-N
Synonym 3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one
PubChem CID 10467624
IUPAC Name (3Z)-3-[(4-chlorophenyl)methylidene]-1,3-dihydro-2-benzofuran-1-one
SMILES ClC1=CC=C(\C=C2/OC(=O)C3=CC=CC=C23)C=C1

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 256.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Chlorobenzal)phthalide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.