accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205437-64-5

IUPAC Name: 2-(4,5-dihydro-1H-imidazol-2-yl)quinoline hydrochloride
Synonyms: 2-(4,5-dihydro-1H-imidazol-2-yl)quinoline hydrochloride
Molecular Weight (g/mol): 233.7
Molecular Formula: C12H12ClN3
InChi Key: DDFHQXAQWZWRSQ-UHFFFAOYSA-N
SMILES: Cl.C1CN=C(N1)C1=CC=C2C=CC=CC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results