accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)(R)OHIOO

CAS RN 205503-96-4

IUPAC Name: (5R,6R)-5,6-dihydroxy-4-iodocyclohexa-1,3-diene-1-carboxylate
Synonyms: (5R,6R)-5,6-dihydroxy-4-iodocyclohexa-1,3-diene-1-carboxylate
Molecular Weight (g/mol): 281.03
Molecular Formula: C7H6IO4
InChi Key: OJVUEUHUFZSXFM-RITPCOANSA-M
SMILES: O[C@@H]1[C@H](O)C(=CC=C1I)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results