CAS RN 205526-28-9
CAS RN 205526-28-9
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[3-(trifluoromethyl)phenyl]propanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[3-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol):
455.43
Molecular Formula:
C25H20F3NO4
InChi Key:
AJMRBDCOLBEYRZ-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC(=CC=C1)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Phe 2.4-dichloro -OH, Advanced ChemTech
CAS: 205526-28-9 Molecular Formula: C25H20F3NO4 Molecular Weight (g/mol): 455.43 InChI Key: AJMRBDCOLBEYRZ-UHFFFAOYNA-N IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[3-(trifluoromethyl)phenyl]propanoic acid SMILES: OC(=O)C(CC1=CC(=CC=C1)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12