accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNIOHO

CAS RN 205526-29-0

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Molecular Weight (g/mol): 513.33
Molecular Formula: C24H20INO4
InChi Key: LXOXXTQKKRJNNB-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)