Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-4-iodo-D-phenylalanine, 98%

Catalog No. AAH6329206
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6329206 5 g
AAH6329203 1 g
AAH63292MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6329206 Supplier Thermo Scientific Chemicals Supplier No. H6329206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 205526-29-0 | C24H20INO4 | 513.33 g/mol

N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205526-29-0
Molecular Formula C24H20INO4
Molecular Weight (g/mol) 513.33
MDL Number MFCD00672557
InChI Key LXOXXTQKKRJNNB-UHFFFAOYNA-N
Synonym fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
PubChem CID 16213204
SMILES OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Beilstein 9159551
Solubility Information Sparingly Soluble in water (6.7E-4g/L) (25°C).
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
Quantity 5 g
Formula Weight 513.32
Sensitivity Light sensitive
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-4-iodo-D-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.