accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)CH3O(S)(S)OOH2C(S)(Z)

CAS RN 20554-84-1

IUPAC Name: (1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one
Synonyms: parthenolide
Molecular Weight (g/mol): 248.32
Molecular Formula: C15H20O3
InChi Key: KTEXNACQROZXEV-QLIGOWBFSA-N
SMILES: C\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)