Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Parthenolide, Tanacetum parthenium

Catalog No. 51273250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-273-250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-273-250MG Supplier MilliporeSigma™ Supplier No. 51273250MG
Add to Cart
Add to Cart

A cell-permeable sesquiterpene lactone with anti-inflammatory, antisecretory, and spasmolytic properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by TLC
Target Proteins the expression of COX-2 and proinflammatory cytokines (TNF-α and IL-1) in LPS-stimulated macrophages
CAS 20554-84-1
Molecular Formula C15H20O3
Formula Weight 248.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.