accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNBrCl

CAS RN 205672-19-1

IUPAC Name: 4-(benzyloxy)-5-bromo-2-chloropyrimidine
Synonyms: 4-(benzyloxy)-5-bromo-2-chloropyrimidine
Molecular Weight (g/mol): 299.55
Molecular Formula: C11H8BrClN2O
InChi Key: PFOACTPILUWRCT-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Br)C(OCC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results