CAS RN 205692-64-4
IUPAC Name:
(3S,5S)-3-(methoxycarbonyl)-5-methyl-4-oxopiperidin-1-ium
Synonyms:
(3S,5S)-3-(methoxycarbonyl)-5-methyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol):
172.2
Molecular Formula:
C8H14NO3
InChi Key:
JIUARERZPGRYNZ-WDSKDSINSA-O
SMILES:
COC(=O)[C@H]1C[NH2+]C[C@H](C)C1=O
1
–
1
of
1
results
1
3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride, 99%
CAS: 205692-64-4 Molecular Formula: C8H13NO3·HCl Molecular Weight (g/mol): 207.66 MDL Number: MFCD00143176 InChI Key: JTZAAEGXSGIXFC-UHFFFAOYSA-N Synonym: methyl 5-methyl-4-oxopiperidine-3-carboxylate hydrochloride,3-methyl-5-methoxycarbonyl-4-piperidone hydrochloride PubChem CID: 73012647 IUPAC Name: methyl 5-methyl-4-oxopiperidine-3-carboxylate;hydrochloride SMILES: CC1CNCC(C1=O)C(=O)OC.Cl