Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride, 99%

Catalog No. AC268085000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268085000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268085000 Supplier Thermo Scientific Chemicals Supplier No. 268085000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 205692-64-4 | C8H13NO3·HCl | 207.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205692-64-4
Molecular Formula C8H13NO3·HCl
Molecular Weight (g/mol) 207.66
MDL Number MFCD00143176
InChI Key JTZAAEGXSGIXFC-UHFFFAOYSA-N
Synonym methyl 5-methyl-4-oxopiperidine-3-carboxylate hydrochloride, 3-methyl-5-methoxycarbonyl-4-piperidone hydrochloride
PubChem CID 73012647
IUPAC Name methyl 5-methyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CC1CNCC(C1=O)C(=O)OC.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 160°C to 175°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 207.66
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.