accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)NH(S)H3CO

CAS RN 205692-65-5

IUPAC Name: (3S,5S)-1-benzyl-3-(methoxycarbonyl)-5-methyl-4-oxopiperidin-1-ium
Synonyms: (3S,5S)-1-benzyl-3-(methoxycarbonyl)-5-methyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol): 262.33
Molecular Formula: C15H20NO3
InChi Key: IVBZCMSOGWPACW-AAEUAGOBSA-O
SMILES: COC(=O)[C@H]1C[NH+](CC2=CC=CC=C2)C[C@H](C)C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results