Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, 99%, Thermo Scientific™

Catalog No. AC268090010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268090010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268090010 Supplier Thermo Scientific Chemicals Supplier No. 268090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 205692-65-5
Molecular Formula C15H19NO3·HCl
Molecular Weight (g/mol) 297.77
MDL Number MFCD00143175
InChI Key UQGAOMLRURDKSU-UHFFFAOYSA-N
Synonym 3-methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, methyl 1-benzyl-5-methyl-4-oxopiperidine-3-carboxylate hydrochloride
PubChem CID 71650946
IUPAC Name methyl 1-benzyl-5-methyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CC1CN(CC(C1=O)C(=O)OC)CC2=CC=CC=C2.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 190°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 297.77
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.