accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHNOONOO(E)

CAS RN 2057-84-3

IUPAC Name: (2E)-1-(2,4-dinitrophenyl)-2-pentylidenehydrazine
Synonyms: (2E)-1-(2,4-dinitrophenyl)-2-pentylidenehydrazine
Molecular Weight (g/mol): 266.26
Molecular Formula: C11H14N4O4
InChi Key: XGVOZGLOQUHZBZ-KPKJPENVSA-N
SMILES: CCCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)