accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20577-26-8

BrNN

CAS RN 20577-26-8

IUPAC Name: 2-bromopyridine-3-carbonitrile
Synonyms: 2-bromopyridine-3-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: QIPXEPRGYVAQFI-UHFFFAOYSA-N
SMILES: BrC1=NC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results