accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHN

CAS RN 20577-27-9

IUPAC Name: 2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 120.11
Molecular Formula: C6H4N2O
InChi Key: DYUMBFTYRJMAFK-UHFFFAOYSA-N
SMILES: O=C1NC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results