accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOOOCH3

CAS RN 20587-30-8

IUPAC Name: methyl 5-methyl-2-nitrobenzoate
Synonyms: methyl 5-methyl-2-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: KFOICDVZQKFCGM-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CC(C)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results