Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Methyl-2-nitrobenzoate 98.0+%, TCI America™
SDP

Catalog No. M25445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25445G 5 g
M254425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25445G Supplier TCI America Supplier No. M25445G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20587-30-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00157275
InChI Key KFOICDVZQKFCGM-UHFFFAOYSA-N
Synonym 5-Methyl-2-nitrobenzoic Acid Methyl Ester, Methyl 6-Nitro-m-toluate, 6-Nitro-m-toluic Acid Methyl Ester
PubChem CID 3314122
IUPAC Name methyl 5-methyl-2-nitrobenzoate
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Methyl-2-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.