accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205985-91-7

IUPAC Name: (2S,4S)-2-methyl-1,3-thiazolidin-3-ium-4-carboxylate
Synonyms: (2S,4S)-2-methyl-1,3-thiazolidin-3-ium-4-carboxylate
Molecular Weight (g/mol): 147.19
Molecular Formula: C5H9NO2S
InChi Key: FHTPNEYXMGZOSH-IUYQGCFVSA-N
SMILES: C[C@H]1[NH2+][C@H](CS1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results