Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4S,2RS)-2-Methylthiazolidine-4-carboxylicacid 97%, Thermo Scientific™

Catalog No. AC292870025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC292870025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292870025 Supplier Thermo Scientific Chemicals Supplier No. 292870025

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205985-91-7
Molecular Formula C5H9NO2S
Molecular Weight (g/mol) 147.192
InChI Key FHTPNEYXMGZOSH-SRBOSORUSA-N
Synonym 4s-2-methyl-1,3-thiazolidine-4-carboxylic acid, 4s,2rs-2-methylthiazolidine-4-carboxylic acid, 4s-2-methylthiazolidine-4-carboxylic acid, 4s,2rs-2-methyl thiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-methyl-, 4s-9ci
PubChem CID 2724963
IUPAC Name (4S)-2-methyl-1,3-thiazolidine-4-carboxylic acid
SMILES CC1NC(CS1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 185°C
Color White
Infrared Spectrum Authentic
Quantity 2.5 g
Formula Weight 147.2
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material (4S, 2RS)-2-Methylthiazolidine-4-carboxylicacid, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.