CAS RN 20601-38-1
IUPAC Name:
[1,1'-bi(cyclohexane)]-4,4'-diol
Synonyms:
[1,1'-bi(cyclohexane)]-4,4'-diol
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
MZXNOAWIRQFYDB-UHFFFAOYSA-N
SMILES:
OC1CCC(CC1)C1CCC(O)CC1
1
–
1
of
1
results
1
4,4'-Bicyclohexanol (mixture of isomers) 98.0+%, TCI America™
CAS: 20601-38-1 Molecular Formula: C12H22O2 Molecular Weight (g/mol): 198.306 MDL Number: MFCD00012056 InChI Key: MZXNOAWIRQFYDB-UHFFFAOYSA-N PubChem CID: 88608 IUPAC Name: 4-(4-hydroxycyclohexyl)cyclohexan-1-ol SMILES: C1CC(CCC1C2CCC(CC2)O)O