Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bicyclohexanol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. B1621500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1621500G 500 g
B162125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1621500G Supplier TCI America Supplier No. B1621500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20601-38-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00012056
InChI Key MZXNOAWIRQFYDB-UHFFFAOYSA-N
PubChem CID 88608
IUPAC Name 4-(4-hydroxycyclohexyl)cyclohexan-1-ol
SMILES C1CC(CCC1C2CCC(CC2)O)O

Specifications

Color White
Quantity 500 g
Synonym 1,1′-Bicyclohexyl-4,4′-diol, 4,4′-Dihydroxydicyclohexyl
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bicyclohexanol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.