accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 206060-54-0

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid
Molecular Weight (g/mol): 431.49
Molecular Formula: C26H25NO5
InChi Key: RHSSAHKTHNKPGU-DEOSSOPVSA-N
SMILES: CC1=CC(O)=CC(C)=C1C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)