CAS RN 20624-25-3
Sodium diethyldithiocarbamate trihydrate, 98%
CAS: 20624-25-3 Molecular Formula: C5H16NNaO3S2 Molecular Weight (g/mol): 225.297 MDL Number: MFCD00150617 InChI Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M Synonym: sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech PubChem CID: 517546 IUPAC Name: sodium;N,N-diethylcarbamodithioate;trihydrate SMILES: CCN(CC)C(=S)[S-].O.O.O.[Na+]
Sodium Diethyldithiocarbamate, Trihydrate, Technical, Spectrum™ Chemical
CAS: 20624-25-3
Sodium diethyldithiocarbamate trihydrate, ACS reagent, ≥99.0%, MilliporeSigma™ Supelco™
MDL Number: MFCD00150617 Synonym: Cupral; Diethyldithiocarbamic acid sodium salt
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
CAS: 20624-25-3 Molecular Formula: C5H10NNaS2·3H2O Molecular Weight (g/mol): 225.31 MDL Number: MFCD00150617 InChI Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M Synonym: sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech PubChem CID: 517546 IUPAC Name: sodium;N,N-diethylcarbamodithioate;trihydrate SMILES: CCN(CC)C(=S)[S-].O.O.O.[Na+]
Sodium diethyldithiocarbamate trihydrate, ACS
CAS: 20624-25-3 Molecular Formula: C5H16NNaO3S2 Molecular Weight (g/mol): 225.297 MDL Number: MFCD00150617 InChI Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M Synonym: sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech PubChem CID: 517546 IUPAC Name: sodium;N,N-diethylcarbamodithioate;trihydrate SMILES: CCN(CC)C(=S)[S-].O.O.O.[Na+]
Sodium Diethyldithiocarbamate, Certified, 0.1% (w/v), LabChem™
CAS: 20624-25-3 Molecular Formula: C5H16NNaO3S2 Molecular Weight (g/mol): 225.297 InChI Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M Synonym: sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech PubChem CID: 517546 IUPAC Name: sodium;N,N-diethylcarbamodithioate;trihydrate SMILES: CCN(CC)C(=S)[S-].O.O.O.[Na+]
Sigma Aldrich Sodium Diethyldithiocarbamate Trihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sodium Diethyldithiocarbamate Trihydrate (Crystalline/Certified ACS), Fisher Chemical
CAS: 20624-25-3 Molecular Formula: C5H16NNaO3S2 Molecular Weight (g/mol): 225.297 MDL Number: MFCD00150617 InChI Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M Synonym: sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech PubChem CID: 517546 IUPAC Name: sodium;N,N-diethylcarbamodithioate;trihydrate SMILES: CCN(CC)C(=S)[S-].O.O.O.[Na+]