Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium diethyldithiocarbamate trihydrate, ACS reagent, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018982 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8982 100 g
11-101-8177 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8982 Supplier MilliporeSigma Supelco Supplier No. 71480100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium diethyldithiocarbamate trihydrate is a chelating agent, generally used in mobilizing toxic metals from the human tissues and experimental animals. It exists as a white or colorless solid, and is freely soluble with gradual decomposition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00150617
Synonym Cupral; Diethyldithiocarbamic acid sodium salt

Specifications

Melting Point 95°C to 98.5°C
CAS 20624-25-3
Physical Form Powder
Quantity 100 g
Linear Formula (C2H5)2 NCSSNa · 3H2 O
Molecular Weight (g/mol) 225.31
Formula Weight 225.31
Percent Purity ≥99.0%
Grade ACS Reagent
Density Vapor Density: 5.9 (vs. air)
Chemical Name or Material Sodium Diethyldithiocarbamate Trihydrate
Show More Show Less
RTECSNumber EZ6550000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.