accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ON

CAS RN 206261-63-4

IUPAC Name: 2-[2-(dimethylamino)ethoxy]benzonitrile
Synonyms: 2-[2-(dimethylamino)ethoxy]benzonitrile
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: DFZYVQTZWFEMTN-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results