accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 206559-61-7

IUPAC Name: 7-ethoxy-1-benzofuran-2-carboxylate
Synonyms: 7-ethoxy-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 205.19
Molecular Formula: C11H9O4
InChi Key: DQJVZIUEXPTEKX-UHFFFAOYSA-M
SMILES: CCOC1=C2OC(=CC2=CC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results