accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3

CAS RN 20665-85-4

IUPAC Name: 4-formyl-2-methoxyphenyl 2-methylpropanoate
Synonyms: 4-formyl-2-methoxyphenyl 2-methylpropanoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: BGKAKRUFBSTALK-UHFFFAOYSA-N
SMILES: COC1=C(OC(=O)C(C)C)C=CC(C=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results