accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHSOOONH4H2O

CAS RN 206659-00-9

IUPAC Name: ammonium 8-(phenylamino)naphthalene-1-sulfonate hydrate
Synonyms: ammonium 8-(phenylamino)naphthalene-1-sulfonate hydrate
Molecular Weight (g/mol): 334.39
Molecular Formula: C16H18N2O4S
InChi Key: WCSOAGYLOWUTAG-UHFFFAOYSA-N
SMILES: [NH4+].O.[O-]S(=O)(=O)C1=CC=CC2=CC=CC(NC3=CC=CC=C3)=C12
Molecular Weight (g/mol) 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results